Anti-TSHB Rabbit Polyclonal Antibody
Catalog # PRSI22-480
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Thyrotropin subunit beta
- Antigen symbol:TSHB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:7252
- Antigen synonyms:Thyrotropin beta chain|Thyrotropin alfa|TSHB|Thyrotropin subunit beta|TSH-beta|TSH-B|Thyroid-stimulating hormone subunit beta
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 21-138 of human TSHB (NP_000540.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: TSHB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse