Anti-KIR2DS4 Rabbit Polyclonal Antibody
PRSI18-099
- Antibody type:Primary
- Antigen name:Killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 4
- Antigen symbol:KIR2DS4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:3809
- Antigen synonyms:p58 NK receptor CL-39/CL-17|NKAT8|killer cell immunoglobulin-like receptor 2DS4|two domains|killer inhibitory receptor 4-1-2|KIR antigen 2DS4|natural killer-associated transcript 8|short cytoplasmic tail|KKA3|NKAT-8|KIR1D|P58 natural killer cell receptor clones CL-39/CL-17|killer cell immunoglobulin-like receptor|4|CD158I|KIR412|p50 killer cell activating receptor KAR-K1d|KIR-2DS4|CD158 antigen-like family member I
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 22-245 of human KIR2DS4 (NP_036446,3)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: KIR2DS4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human