Order Entry
France
ContactUsLinkComponent
 
Anti-PRKAA1/PRKAA2 Rabbit Polyclonal Antibody
  PRSI16-906
New Product
Anti-PRKAA1/PRKAA2 Rabbit Polyclonal Antibody
  PRSI16-906
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    Protein kinase, AMP-activated, alpha 1 catalytic subunit
  • Antigen symbol:
    PRKAA1/PRKAA2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    5562;5563
  • Antigen synonyms:
    AMPK|HMG-CoA redustase kinase|VEGFR2|AMPK-alpha1|PRKAA1|AAPK1|AMPKa1|AMPK alpha-1 chain|FLK1
  • Modification:
    None
  • Storage buffer:
    PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
  • Storage temperature:
    Store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    Batch dependent
  • Shipping temperature:
    20
  • Immunogen:
    Recombinant protein of human PRKAA1/PRKAA2
  • Purification:
    Affinity purification
  • Pack type:
    Vial
  • Pk:
    50 µl

 

 

Type: Primary
Antigen: PRKAA1/PRKAA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat