Anti-DOCK4 Rabbit Polyclonal Antibody
Catalog # PRSI16-220
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Dedicator of cytokinesis 4
- Antigen symbol:DOCK4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:9732
- Antigen synonyms:dedicator of cytokinesis protein 4
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 150-450 of human DOCK4 (NP_055520,3)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: DOCK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human