Order Entry
France
ContactUsLinkComponent
Anti-CCAR1 Rabbit Polyclonal Antibody
Anti-CCAR1 Rabbit Polyclonal Antibody
Catalog # PRSI22-137
Supplier:  ProSci Inc.
New Product
Anti-CCAR1 Rabbit Polyclonal Antibody
Catalog # PRSI22-137
Supplier:  ProSci Inc.
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cell division cycle and apoptosis regulator 1
  • Antigen symbol:
    CCAR1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    55749
  • Antigen synonyms:
    cell division cycle and apoptosis regulator 1|MGC44628|CARP1|OTTHUMP00000019709|CARP-1|DIS|cell-cycle and apoptosis regulatory protein 1|RP11-437A18.1|CCAR1
  • Storage buffer:
    PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
  • Storage temperature:
    Store at −20...+730 °C. Avoid freeze/thaw cycles.
  • Concentration:
    Batch dependent
  • Shipping temperature:
    20
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human CCAR1 (NP_060707.2).
  • Purification:
    Affinity purification
  • Pack type:
    Vial
  • Pk:
    50 µl

Specifications

About this item

Type: Primary
Antigen: CCAR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human