Anti-HMG20B Rabbit Polyclonal Antibody
Catalog # PRSI19-360
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:High mobility group 20B
- Antigen symbol:HMG20B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:10362
- Antigen synonyms:HMG domain-containing protein HMGX2|Sox-like transcriptional factor|SMARCE1-related protein|BRCA2-associated factor 35|BRAF25|SOXL|pp8857|SMARCE1r|HMG box domain containing 2|PP7706|member 1-related|BRAF35|HMG box-containing protein 20B|structural DNA-binding protein BRAF35|SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily E|SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily E member 1-related|HMGXB2|HMG domain-containing protein 2|HMGX2
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-317 of human HMG20B (NP_006330.2).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: HMG20B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat