Anti-DOCK2 Rabbit Polyclonal Antibody
Catalog # PRSI19-099
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:Dedicator of cytokinesis 2
- Antigen symbol:DOCK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:1794
- Antigen synonyms:IMD4dedicator of cytokinesis protein 2|dedicator of cyto-kinesis 2
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1551-1830 of human DOCK2 (NP_004937.1).
- Purification:Affinity purification
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: DOCK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse