Order Entry
France
ContactUsLinkComponent
Anti-SLC1A1 Rabbit Polyclonal Antibody
Anti-SLC1A1 Rabbit Polyclonal Antibody
  PRSI18-862
New Product
Anti-SLC1A1 Rabbit Polyclonal Antibody
  PRSI18-862
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    Solute carrier family 1 member 1
  • Antigen symbol:
    SLC1A1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    6505
  • Antigen synonyms:
    DCBXA|neuronal and epithelial glutamate transporter|system Xag)|excitatory amino acid transporter 3|sodium-dependent glutamate/aspartate transporter 3|SCZD18|member 1|EAAC1|solute carrier family 1 (neuronal/epithelial high affinity glutamate transporter|excitatory amino acid carrier 1|EAAT3
  • Storage buffer:
    PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
  • Storage temperature:
    Store at −20...+730 °C. Avoid freeze/thaw cycles.
  • Concentration:
    Batch dependent
  • Shipping temperature:
    20
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 430-524 of human SLC1A1 (NP_004161.4).
  • Purification:
    Affinity purification
  • Pack type:
    Vial
  • Pk:
    50 µl

 

 

Type: Primary
Antigen: SLC1A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat