Anti-SMCP Rabbit Polyclonal Antibody
Catalog # PRSI56-673
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Sperm mitochondrial-associated cysteine-rich protein
- Antigen symbol:SMCP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:Amino acid range = 68-97
- Form:Liquid
- Gene ID:P49901
- Modification:None
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide
- Molecular weight:13 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This SMCP antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 68-97 amino acids from the C-terminal region of human SMCP
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
Specifications
About this item
Sperm mitochondria differ in morphology and subcellular localization from those of somatic cells. They are elongated, flattened, and arranged circumferentially to form a helical coiled sheath in the midpiece of the sperm flagellum. The protein encoded by this gene localizes to the capsule associated with the mitochondrial outer membranes and is thought to function in the organization and stabilization of the helical structure of the sperm's mitochondrial sheath.
- For WB starting dilution is: 1:1000
Type: Primary
Antigen: SMCP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human