Anti-FCER1A Hamster Monoclonal Antibody [clone: FceR1] (FITC (Fluorescein Isothiocyanate))
Catalog # PRSI77-177
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide
- Antigen symbol:FCER1A
- Clonality:Monoclonal
- Clone:FceR1
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Hamster
- Isotype:IgG
- Reactivity:Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:P12319
- Antigen synonyms:FCER1A
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:0,025 mg
Specifications
About this item
The Mar-1 monoclonal antibody binds to the Fc epsilon Receptor I alpha subunit (FceR1a), which is a transmembrane glycoprotein from the immunoglobulin superfamily. FceR1a lacks signal-transducing ability and is expressed by mast and basophil cells.The Fc epsilon Receptor I alpha subunit is upregulated by IgE and forms a tetramer with a beta subunit and two gamma subunits, which have ITAM (immunoreceptor tyrosine-based activation motifs). The complex formed by the four subunits has very important roles in the IgE-facilitated allergic reactions.
Type: Primary
Antigen: FCER1A
Clonality: Monoclonal
Clone: FceR1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse