Order Entry
France
ContactUsLinkComponent
Anti-Cd69 Hamster Monoclonal Antibody [clone: H1.2F3]
Anti-Cd69 Hamster Monoclonal Antibody [clone: H1.2F3]
Catalog # PRSI76-817
Supplier:  ProSci Inc.
Anti-Cd69 Hamster Monoclonal Antibody [clone: H1.2F3]
Catalog # PRSI76-817
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD69 antigen
  • Antigen symbol:
    Cd69
  • Clonality:
    Monoclonal
  • Clone:
    H1.2F3
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Hamster
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Cross adsorption:
    No
  • Format:
    purified antibody
  • Form:
    liquid
  • Gene ID:
    P37217
  • Antigen synonyms:
    Cd69
  • Storage buffer:
    Phosphate-buffered aqueous solution, ph 7,2.
  • Storage temperature:
    The product should be stored undiluted at 4 °C. Do not freeze.
  • Shipping temperature:
    4 °C
  • Tested applications:
    Functional
  • Purification:
    The monoclonal antibody was purified utilizing affinity chromatography. The endotoxin level is determined by LAL test to be less than 0,01 EU/µg of the protein.
  • Pk:
    0,1 mg

Specifications

About this item

The H1.2F3 monoclonal antibody specifically reacts with human CD69, the 27 to 33 kDa type II transmembrane protein also known as the very early activation antigen (VEA) or the activation inducer molecule (AIM). It is expressed as a disulfide-linked dimer on B cells, T cells, NK cells, platelets, eosinophils, and neutrophils. It increases in expression upon cell activation and seems to serve a role as a signaling receptor.

Type: Primary
Antigen: Cd69
Clonality: Monoclonal
Clone: H1.2F3
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse