Order Entry
France
ContactUsLinkComponent
Anti-SELP Mouse Monoclonal Antibody [clone: Psel.KO2.3] (PE (Phycoerythrin))
Anti-SELP Mouse Monoclonal Antibody [clone: Psel.KO2.3] (PE (Phycoerythrin))
Catalog # PRSI76-209
Supplier:  ProSci Inc.
New Product
Anti-SELP Mouse Monoclonal Antibody [clone: Psel.KO2.3] (PE (Phycoerythrin))
Catalog # PRSI76-209
Supplier:  ProSci Inc.
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    selectin P (granule membrane protein 140kDa, antigen CD62)
  • Antigen symbol:
    SELP
  • Clonality:
    Monoclonal
  • Clone:
    Psel.KO2.3
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    P16109; P18337
  • Antigen synonyms:
    SELP
  • Storage buffer:
    Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
  • Storage temperature:
    The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
  • Shipping temperature:
    4 °C
  • Purification:
    The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
  • Pk:
    0,1 mg

Specifications

About this item

The Psel.KO2.3 monoclonal antibody specifically reacts with mouse CD62P (P-Selectin), a 140 kDa type I transmembrane glycoprotein also known as granule membrane protein 140 (GMP-140). Upon activation it is quickly transported to the plasma membrane from the alpha-granules of platelets and WEibel-Palade bodies of endothelial cells. It is essential to the functions of cell adhesion during inflammatory reactions and in the interaction of platelets with monocytes and neutrophils. The Psel.KO2.3 antibody does not block adhesion and binds to an epitope common to several species including mouse, human, and rat.

Type: Primary
Antigen: SELP
Clonality: Monoclonal
Clone: Psel.KO2.3
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Mouse