Anti-CD63 Recombinant Antibody [Clone: MOF11]
Catalog # PRSI24-358
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Antigen name:CD63 molecule
- Antigen symbol:CD63
- Clonality:Recombinant
- Clone:MOF11
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Isotype:IgG1 kappa
- Cross adsorption:No
- Form:Liquid
- Gene ID:967
- Antigen synonyms:OMA81H|ME491|Granulophysin|LAMP-3|LAMP3|MLA1|TSPAN30
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:Human monocytes
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: CD63 molecule
Clonality: Recombinant
Clone: MOF11
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1 kappa
Reactivity: Human