Anti-CD22 Recombinant Antibody [Clone: hL22 (Epratuzumab)]
PRSI24-491
- Antibody type:Primary
- Antigen name:CD22 molecule
- Antigen symbol:CD22
- Clonality:Recombinant
- Clone:hL22 (Epratuzumab)
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Cynomolgus monkey,Rhesus monkey,Human
- Cross adsorption:No
- Form:Liquid
- Gene ID:933
- Antigen synonyms:B-lymphocyte cell adhesion molecule|B-cell receptor CD22|Sialic acid-binding Ig-like lectin 2|BL-CAM|Siglec-2|Leu14|T-cell surface antigen Leu-14
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody was prepared by the humanization of LL2 (EPB-2), a murine anti-CD22 IgG2a raised against Raji Burkitt lymphoma cells. Murine sequences comprise 5–10% of the molecule, with the remainder being human framework sequences, which greatly reduces the potential for immunogenicity (Traczewski, 2010)
- Tested applications:Blocking
- Pack type:Vial
- Pk:0,2 mg
Type: Primary,Biosimilars
Antigen: CD22 molecule
Clonality: Recombinant
Clone: hL22 (Epratuzumab)
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG1 kappa
Reactivity: Human;Rhesus Monkey;Cynomolgus Monkey