Order Entry
France
ContactUsLinkComponent
Bovine trypsin (from pancreas), MP Biomedicals
Bovine trypsin (from pancreas), MP Biomedicals
Supplier:  MP Biomedicals
Certificates
Bovine trypsin (from pancreas), MP Biomedicals
Supplier:  MP Biomedicals
Certificates

Specifications

  • CAS No.:
    9002-07-7
  • Source:
    Pancreas
  • Species:
    Bovine
  • Storage conditions:
    +4 °C
  • Form:
    White lyophilized powder
  • MDL No.:
    MFCD00082094
  • Enzyme name:
    Trypsin
  • Enzyme activity:
    ≥180 TAME u/mg
  • Purity:
    2X crystallized
  • pH:
    8,0

Specifications

Related Information

Frequently Bought Together