Order Entry
France
ContactUsLinkComponent
Riboflavine (vitamin B2)
Riboflavine (vitamin B2)
Catalog # 47861.
Supplier:  Merck
CAS Number:  
undefined
Riboflavine (vitamin B2)
Catalog # 47861.
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :376.37 g/mol
Merck Index:12,08367
Melting Pt:280 °C
Storage Temperature:Ambient
EINECS:201-507-1
CAS Number:83-88-5
Formula:C₁₇H₂₀N₄O₆
MDL Number:MFCD00005022