Order Entry
France
ContactUsLinkComponent
Riboflavine (vitamin B2), Millipore®
Riboflavine (vitamin B2), Millipore®
Catalog # 555682-10
CAS Number:  
Riboflavine (vitamin B2), Millipore®
Catalog # 555682-10
CAS Number:  

Specifications

  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :376.37 g/mol
Merck Index:12,08367
Melting Pt:280 °C
Storage Temperature:Ambient
EINECS:201-507-1
CAS Number:83-88-5
Formula:C₁₇H₂₀N₄O₆
MDL Number:MFCD00005022