Riboflavine (vitamin B2) ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:83-88-5
:
Formula:C₁₇H₂₀N₄O₆
MDL Number:MFCD00005022
:376.37 g/mol
Merck Index:14,8200
Melting Pt:290 °C
Storage Temperature:Refrigerator
EINECS:201-507-1
Related Information
Certificates
Search by Lot NumberALFAA11764.14_TDS_EN ALFAA11764.22_TDS_EN
Product specification
Organic Building Blocks and Intermediates
Application Notes
Frequently Bought Together