Order Entry
France
ContactUsLinkComponent
Phenol red sodium salt dye pH-indicator, Sigma-Aldrich®
Irritant
Phenol red sodium salt dye pH-indicator, Sigma-Aldrich®
Certificates
undefined
Irritant
Phenol red sodium salt dye pH-indicator, Sigma-Aldrich®

Specification Test Results

  • Appearance (Colour):
    Red to Brown
  • Appearance (Form):
    Powder
  • Infrared spectrum:
    Conforms to Structure
  • Solubility (Colour):
    Dark Red
  • Solubility (Form), 1 mg/ml, H₂O:
    Solution
  • pH Pass:
    6.8 (Yellow) to 8.2 (Red)
  • Recommended Retest Period:
    5 Years

Specification Test Results

Frequently Bought Together