Order Entry
France
ContactUsLinkComponent
Orthoboric acid 40 g/l in aqueous solution
Orthoboric acid 40 g/l in aqueous solution
Catalog # VWRS307868.1000
Supplier:  VWR Collection
CAS Number:  
undefined
Orthoboric acid 40 g/l in aqueous solution
Catalog # VWRS307868.1000
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid
 :
 :61.83 g/mol
Storage Temperature:Ambient
EINECS:233-139-2
CAS Number:10043-35-3
Formula:B(OH)₃
MDL Number:MFCD00011337