Order Entry
France
ContactUsLinkComponent
Methyl orange 0,2% in aqueous solution
Methyl orange 0,2% in aqueous solution
Catalog # PRLS87182.260
Supplier:  PROLABO CMP
CAS Number:  
Methyl orange 0,2% in aqueous solution
Catalog # PRLS87182.260
Supplier:  PROLABO CMP
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulphonic acid, sodium salt, Methyl orange sodium salt, Helianthine
 :
 :327.34 g/mol
Storage Temperature:Ambient
EINECS:208-925-3
CAS Number:547-58-0
Formula:C₁₄H₁₄N₃NaO₃S
MDL Number:MFCD00007502