Order Entry
France
ContactUsLinkComponent
 
Bromocresol green 0.1% in aqueous solution
Catalog # PRLSMC302454.0100
Supplier:  PROLABO CMP
CAS Number:  
undefined
Bromocresol green 0.1% in aqueous solution
Catalog # PRLSMC302454.0100
Supplier:  PROLABO CMP
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :698.02 g/mol
Storage Temperature:Ambient
EINECS:200-972-8
CAS Number:76-60-8
Formula:C₂₁H₁₄Br₄O₅S
MDL Number:MFCD00005874