Order Entry
France
Orders LinkContactUsLinkComponent
Congo red

Congo red

5 g
Catalog # ROTH0322.1
Health
Marketsource
Supplier:   Roth Carl
Congo red
Catalog # ROTH0322.1
Supplier:  Roth Carl
Marketsource
CAS Number:  573-58-0
Congo red

5 g

CAS Number:  573-58-0
Price
73,34 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Disodium 3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalene-1-sulphonate), Sodium 3,3'-([1,1'-biphenyl]-4,4'-diylbis(diazene-2,1-diyl))bis(4-aminonaphthalene-1-sulphonate), CI 22120, Direct Red 28
  • CAS Number:573-58-0
  • Formula:C₃₂H₂₂N₆Na₂O₆S₂
  • MDL Number:MFCD00004028
  • Molecular Weight:696.67 g/mol
  • Merck Index:12,02562
  • Melting Pt:> 360 °C
  • Storage Temperature:Ambient
  • EINECS:209-358-4
Specifications

Specifications

  • Pk:
    5 g

Specifications