Order Entry
France
ContactUsLinkComponent
Mycophenolate mofetil ≥98% (by HPLC), white to off-white solid
Irritant
Environment
Mycophenolate mofetil ≥98% (by HPLC), white to off-white solid
Supplier:  ENZO LIFE SCIENCES
Certificates
undefined
Irritant
Environment
Mycophenolate mofetil ≥98% (by HPLC), white to off-white solid
Supplier:  ENZO LIFE SCIENCES

Specification Test Results

  • Purity:
    ≥98% (HPLC)
  • Appearance:
    White to off-white solid

Specification Test Results

Frequently Bought Together