Order Entry
France
ContactUsLinkComponent
Calcimycin (A23187) (from Streptomyces chartreusis) ≥99% (by TLC)
Calcimycin (A23187) (from Streptomyces chartreusis) ≥99% (by TLC)
Catalog # ENZOBMLCA1000010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Calcimycin (A23187) (from Streptomyces chartreusis) ≥99% (by TLC)
Catalog # ENZOBMLCA1000010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :523.63 g/mol
Storage Temperature:Freezer
CAS Number:52665-69-7
Formula:C₂₉H₃₇N₃O₆
MDL Number:MFCD00151202