Order Entry
France
ContactUsLinkComponent
Tranilast ≥95% (by HPLC)
Tranilast ≥95% (by HPLC)
Catalog # ENZOALX550409M010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Tranilast ≥95% (by HPLC)
Catalog # ENZOALX550409M010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :327.34 g/mol
CAS Number:53902-12-8
Formula:C₁₈H₁₇NO₅
MDL Number:MFCD00864787