

(6R)-5,6,7,8-Tetrahydro-L-biopterin dihydrochloride ≥98% (by HPLC)
Supplier: ENZO LIFE SCIENCES
New Product
Certificates
About this item
Merck Index:14,08368
Specification Test Results
- Purity (HPLC):≥98%
- Appearance:White to off-white powder.
Specification Test Results
Frequently Bought Together