Sodium nitroprusside dihydrate
Catalog #
Product Details & Documents
Synonyms: Sodium pentacyanonitrosylferrate(III) dihydrate, Sodium nitrosopentacyanoferrate (III) dihydrate, Sodium nitroferricyanide(III) dihydrate, Disodium pentacyanonitrosylferrate (III) dihydrate, Sodium nitroferricyanide dihydrate
- CAS Number:13755-38-9
- Formula:Na₂[Fe(CN)₅NO]·2H₂O
- MDL Number:MFCD00149192
- Molecular Weight:297.95 g/mol
- Merck Index:13,08722
- Density:1,72 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:238-373-9
- UN:1588
- ADR:6.1,III



