Skip to Header
Skip to Main Content
Skip to Footer
Avancé
Avancé
Fiches de données de sécurité (FDS)
Certificats
Données du fabricant
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tosufloxacin tosylate
Tosufloxacin tosylate ≥98% (by HPLC)
Tosufloxacin tosylate ≥98% (by HPLC)
ENZOALX380303M250
:
ENZO LIFE SCIENCES
:
Tosufloxacin tosylate ≥98% (by HPLC)
ENZOALX380303M250
:
ENZO LIFE SCIENCES
:
Pk:
250 mg
Seq:
2590
:
:
404.4 · 172.2 g/mol
:
Freezer
:
115964-29-9
:
C₁₉H₁₅F₃N₄O₃·C₇H₈O₃S
:
MFCD01711971