Order Entry
France
ContactUsLinkComponent
Kanamycin B sulphate ≥99% (by HPLC), high purity
Kanamycin B sulphate ≥99% (by HPLC), high purity
Catalog # ENZOALX380095M250
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
Kanamycin B sulphate ≥99% (by HPLC), high purity
Catalog # ENZOALX380095M250
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Seq:
    2590
  • Environmentally Preferable:

Specifications

About this item

 :
 :581.6 g/mol
CAS Number:29701-07-3
Formula:C₁₈H₃₇N₅O₁₀·H₂SO₄