Netropsin dihydrochloride ≥90% (by HPLC)
Catalog # ENZOALX380088M005
Supplier: ENZO LIFE SCIENCES
New Product
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
- Environmentally Preferable:
Specifications
Specification Test Results
- Purity:≥90% (HPLC)
- Source:Isolated from Streptomyces netropsis
Specification Test Results
About this item
:
:503.5 g/mol
:GHS06
Storage Temperature:Freezer
CAS Number:1438-30-8
Formula:C₁₈H₂₆N₁₀O₃·2HCl