Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ketotifen fumarate
Ketotifen fumarate ≥99%
Ketotifen fumarate ≥99%
Ketotifen fumarate ≥99%
ENZOBMLNH1060001
:
ENZO LIFE SCIENCES
:
Ketotifen fumarate ≥99%
ENZOBMLNH1060001
:
ENZO LIFE SCIENCES
:
Pk:
1 g
Seq:
3047
:
:
425.50 g/mol
:
34580-14-8
:
C₁₉H₁₉NOS
:
MFCD00079394