Order Entry
France
ContactUsLinkComponent
NG,NG'-Dimethyl-L-arginine dihydrochloride ≥98% (by TLC)
NG,NG'-Dimethyl-L-arginine dihydrochloride ≥98% (by TLC)
Catalog # ENZOALX106008M025
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
undefined
NG,NG'-Dimethyl-L-arginine dihydrochloride ≥98% (by TLC)
Catalog # ENZOALX106008M025
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Purity:
    ≥98% (TLC)

Specification Test Results

About this item

Synonyms: SDMA dihydrochloride, SDMA.2HCl, Symmetric dimethylarginine dihydrochloride, H-Arg(di-Me)-OH.2HCl (symmetrical)
 :
 :275.2 g/mol
Storage Temperature:Freezer
CAS Number:30344-00-4
Formula:C₈H₁₈N₄O₂·2HCl