D-Maltotriose, Reference standards
EHERA14734950WA100
:
- Concentration:1000 µg/ml in water
- Pk:1 mL
- CAS No.:1109-28-0
- Description:D-Maltotriose
- Shipping conditions:20 °C
- Formula:C₁₈H₃₂O₁₆
- Storage temperature:4 °C
- Molecular weight (Mw):504,44
- Form:Solution