Heptafluorobutyric anhydride (HFAA) ≥99.0% for GC derivatisation, Supelco®
Catalog # 394912-5ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
Specifications
About this item
:
:410.06 g/mol
Boiling Pt:108…110 °C (lit.)
Melting Pt:–43 °C (lit.)
Density:1.674 g/mL (20 °C) (lit.)
EINECS:206-410-8
UN:3625
ADR:8,II
:M-Clarity™ quality level = MQ100
:GHS05
CAS Number:336-59-4
Formula:C₈F₁₄O₃
MDL Number:MFCD00000432