Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ammonium heptamolybdate solutions
Ammonium heptamolybdate 10% in aqueous solution
Ammonium heptamolybdate 10% in aqueous solution
PRLS85699.290
:
PROLABO CMP
:
Ammonium heptamolybdate 10% in aqueous solution
PRLS85699.290
:
PROLABO CMP
:
Pack type:
Plastic bottle
Pk:
1 L
Seq:
5037
Environmentally Preferable:
:
:
1235.86 g/mol
:
234-722-4
:
12054-85-2
:
(NH₄)₆Mo₇O₂₄·4H₂O
:
MFCD00167059