Ammonium heptamolybdate 10% in aqueous solution
Supplier: PROLABO CMP
Certificates
About this item
CAS Number:12054-85-2
:
Formula:(NH₄)₆Mo₇O₂₄·4H₂O
MDL Number:MFCD00167059
:1235.86 g/mol
EINECS:234-722-4
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together