Order Entry
France
ContactUsLinkComponent
β-Alanine ≥99%, Millipore®
β-Alanine ≥99%, Millipore®
Supplier:  Merck
Certificates
β-Alanine ≥99%, Millipore®
Supplier:  Merck
Certificates

Specification Test Results

  • Assay:
    99% min
  • Heavy metals (as Pb):
    0.001% max
  • Identity (IR-spectrum):
    passes test
  • TLC-test:
    passes test

Specification Test Results

Related Information

Frequently Bought Together