β-Alanine ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:107-95-9
:
Formula:C₃H₇NO₂
MDL Number:MFCD00008200
:89.09 g/mol
Merck Index:14,205
Boiling Pt:201 °C (1013 hPa)
Melting Pt:∼ 205 °C (dec.)
Density:1.437
Storage Temperature:Ambient
EINECS:203-536-5
Related Information
Frequently Bought Together