Anti-SARS-CoV-2 nsp12 Rabbit Monoclonal Antibody [clone: HL1444]
Catalog # NOVUNBP3-25653
Supplier: Novus Biologicals
Specifications
- Antigen name:SARS-CoV-2 nsp12
- Clonality:Monoclonal
- Clone:HL1444
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Virus
- Western blot:Yes
- Format:Azide and BSA free
- Gene ID:ORF1ab
- Antigen synonyms:pp1a|Non-structural protein 12|Replicase polyprotein 1a|ORF1ab polyprotein|nsp12|ORF1a polyprotein
- Purity:Protein A purified.
- Storage buffer:PBS
- Storage temperature:Store at 4 °C short term. Aliquot and store at −20 °C long term. Avoid freeze-thaw cycles.
- Immunogen:Recombinant protein encompassing a sequence within the center region of SARS-CoV-2 nsp12/RdRP. The exact sequence is proprietary.
- Pk:100 µl
Specifications
About this item
- Immunocytochemistry/Immunofluorescence: Assay dependent
- Immunohistochemistry: Assay dependent
- Immunohistochemistry-Paraffin: Assay dependent
- Western blot: 1:1000 - 1:10000
Caution: Research use only.
Type: Primary
Antigen: SARS-CoV-2 nsp12
Clonality: Monoclonal
Clone: HL1444
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus