Anti-Human TOP1 Rabbit Polyclonal Antibody
USBI337594
- Antibody type:Primary
- Antigen name:Topoisomerase (DNA) I
- Antigen symbol:TOP1
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:P11387
- Antigen synonyms:TOP1|TOPI
- Purity:Purified by immunoaffinity chromatography.
- Storage temperature:Store at –20 °C
- Tested applications:Suitable for use in ELISA and Western Blot. Other applications not tested
- Size:100 μl
- Pk:100 µl
Type: Primary
Antigen: Human TOP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human