Order Entry
France
Orders LinkContactUsLinkComponent
Table Top Refrigerators
 :   Liebherr
Table Top Refrigerators
 :  Liebherr
Table Top Refrigerators
Table-top refrigerators for general use.
  • Very cool
  • Interior LED lighting
  • TouchControl
  • Durable work surface
  • Slim handle equipped with an integrated opening mechanism
  • Dishwasher safe equipment
  • Satin glass shelves with decorative stainless steel strips
These refrigerators comes with screen lock, door alarm, height adjustable door shelves, unclipable door seal, reversible door stop, egg tray and vegetable drawers.
  • Catalog No:
  • LIEBKTE551
  • LIEBKTE550
  • LIEBRE1000
  • Temperature:
  • 3...9 °C
  • 3...9 °C
  • 3...9 °C
  • Defrost:
  • Automatic
  • Automatic
  • Automatic
  • Shelves/drawers:
  • 2
  • 4
  • 2
  • Material:
  • Steel
  • Steel
  • Steel
  • Sound level:
  • 37 dB
  • 35 dB
  • 35 dB
  • Energy class:
  • E
  • E
  • E
  • Electrical:
  • 220 - 240 V, 50 Hz, 1,2 A
  • 220 - 240 V, 50 Hz, 1,2 A
  • 220 - 240 V, 50 Hz, 1,2 A
  • Weight:
  • 34,1 kg
  • 34 kg
Product Family Options
KTE model
  • Description
    Capacity
    W×D×H ext.
  • Table top refrigerator, KTE551
    111 L
    550×607×850 mm
    1 * 1 Pce
    1 * 1 Pce - 537,03 €
    537,03 €
    1 * 1 Pce
       537,03 €
    Specifications
    Cat. No.
    LIEBKTE551
    Temperature
    3...9 °C
    Defrost
    Automatic
    Shelves/drawers
    2
    Material
    Steel
    Sound level
    37 dB
    Energy class
    E
    Electrical
    220 - 240 V, 50 Hz, 1,2 A
    Weight
    34,1 kg
  • Table top refrigerator, KTE550
    125 L
    550×607×850 mm
    1 * 1 Pce
    1 * 1 Pce - 488,13 €
    488,13 €
    1 * 1 Pce
       488,13 €
    Specifications
    Cat. No.
    LIEBKTE550
    Temperature
    3...9 °C
    Defrost
    Automatic
    Shelves/drawers
    4
    Material
    Steel
    Sound level
    35 dB
    Energy class
    E
    Electrical
    220 - 240 V, 50 Hz, 1,2 A
    Weight
    34 kg
RE model
  • Description
    Capacity
    W×D×H ext.
  • Compact table top refrigerator, RE1000
    92 L
    550×607×850 mm
    1 * 1 Pce
    1 * 1 Pce - 615,30 €
    615,30 €
    1 * 1 Pce
       615,30 €
    Specifications
    Cat. No.
    LIEBRE1000
    Temperature
    3...9 °C
    Defrost
    Automatic
    Shelves/drawers
    2
    Material
    Steel
    Sound level
    35 dB
    Energy class
    E
    Electrical
    220 - 240 V, 50 Hz, 1,2 A
    Weight