Order Entry
France
ContactUsLinkComponent
Charcoal activated, rods, Norit® RBAA-3, Supelco®
Charcoal activated, rods, Norit® RBAA-3, Supelco®
Catalog # 29238-1KG
Supplier:  Merck
CAS Number:  
Charcoal activated, rods, Norit® RBAA-3, Supelco®
Catalog # 29238-1KG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :12.01 g/mol
CAS Number:7440-44-0
 :64365-11-3
Formula:C
MDL Number:MFCD00133992