Silica gel (7734) pore size 60 Å, 70-230 mesh, high purity, Supelco®
Catalog # 403598-500G
Supplier: Merck
CAS Number:
Specifications
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:60.08 g/mol
Boiling Pt:2230 °C
Melting Pt:> 1600 °C
Storage Temperature:Ambient
EINECS:231-545-4
CAS Number:63231-67-4
:7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232