Order Entry
France
ContactUsLinkComponent
Anti-Zika Membrane Rabbit Polyclonal Antibody
Anti-Zika Membrane Rabbit Polyclonal Antibody
Catalog # PRSI8667
Supplier:  ProSci Inc.
Anti-Zika Membrane Rabbit Polyclonal Antibody
Catalog # PRSI8667
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    flavivirus polyprotein
  • Antigen symbol:
    Zika Membrane
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Zika virus (ZIKV)
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    7751225
  • Antigen synonyms:
    Zika M|Zika Membrane Antibody: Zika Mem
  • Modification:
    None
  • Storage buffer:
    Zika membrane antibody is supplied in PBS containing 0,02% sodium azide.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    20
  • Immunogen:
    Zika membrane antibody was raised against a peptide corresponding to 14 amino acids near the amino terminus of human Zika membrane.
  • Purification:
    Zika membrane antibody is affinity chromatography purified via peptide column.
  • Size:
    0,1 mg
  • Pack type:
    Vial
  • Pk:
    0,1 mg

Specifications

About this item

Zika membrane antibody can be stored at 4 to 730 °C for three months and –20 to +730 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.

Type: Primary
Antigen: Zika Membrane
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Zika