Order Entry
France
ContactUsLinkComponent
Silver nitrate 0.1 mol/l (0.1 N) in aqueous solution
Silver nitrate 0.1 mol/l (0.1 N) in aqueous solution
Catalog # FFCHFF61580500
Supplier:  FF CHEMICALS
CAS Number:  
undefined
Silver nitrate 0.1 mol/l (0.1 N) in aqueous solution
Catalog # FFCHFF61580500
Supplier:  FF CHEMICALS
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt
 :
 :169.87 g/mol
EINECS:231-853-9
UN:3082
 :GHS07
 :GHS09
Storage Temperature:Ambient
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414
Boiling Pt:444
Melting Pt:210
ADR:9 III
Merck Index:12,08661