Order Entry
France
ContactUsLinkComponent
Sulphuric acid 1 mol/l (2 N) in aqueous solution
Sulphuric acid 1 mol/l (2 N) in aqueous solution
Catalog # FFCHFF6575000
Supplier:  FF CHEMICALS
CAS Number:  
Sulphuric acid 1 mol/l (2 N) in aqueous solution
Catalog # FFCHFF6575000
Supplier:  FF CHEMICALS
CAS Number:  

Specifications

  • Pk:
    5 L
  • Seq:
    5190
  • Environmentally Preferable:

Specifications

About this item

 :
 :98.08 g/mol
Storage Temperature:Ambient
EINECS:231-639-5
UN:2796
ADR:8,II
 :GHS07
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589