Order Entry
France
ContactUsLinkComponent
 
Hydrochloric acid 3 mol/l (3 N)
Catalog # FFCHFF8260500
Supplier:  FF CHEMICALS
CAS Number:  
Hydrochloric acid 3 mol/l (3 N)
Catalog # FFCHFF8260500
Supplier:  FF CHEMICALS
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Hydrochloric acid solution, Muriatic acid
 :
Density:1.06 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-595-7
ADR:8,II
 :36.46 g/mol
UN:1789
 :GHS07
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324