Hydrochloric acid 33%
Catalog # FFCHFF14615000
Supplier: FF CHEMICALS
CAS Number:
Specifications
- Pk:5 L
- Seq:5190
- Environmentally Preferable:
Specifications
About this item
Synonyms: Hydrochloric acid solution, Muriatic acid
:
:36.46 g/mol
EINECS:231-595-7
UN:1789
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324
Melting Pt:–30 °C
Density:1.18 g/cm³ (20 °C)
ADR:8 II