Hydrochloric acid 200 g/l
Supplier: FF CHEMICALS
Certificates
About this item
Synonyms: Hydrochloric acid solution, Muriatic acid
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Molecular Weight:36.46 g/mol
Storage Temperature:Ambient
EINECS:231-595-7
UN:1789
ADR:8 II
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together